Learn More
VMAT2 Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136170
Catalog No. 01-672-119
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 μL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
Transports N-methyl-4-phenylpyridinium (MPP+), a neurotoxin metabolite associated with Parkinson′s disease [RGD].
Specifications
Specifications
VMAT2
Polyclonal
whole serum with 0.1% sodium azide
Q05940, Q8BRU6, Q01827
MGC120477, MGC120478, MGC26538, SVAT, SVMT, VAT2, VMAT2
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Porcine, Rat
Immunohistochemistry (Frozen)
Unconjugated
SLC18A2
Lyophilized
SLC18A2
Synthetic peptide from the C-terminal of rat VMAT2 conjugated to BSA.
50 μL
4° C
Primary
100154950, 214084, 25549, 6571