Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VMAT2 Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522864

Catalog No. PIPA522864


Add to cart

Description

Description

VMAT2 is involved in the ATP-dependent vesicular transport of biogenic amine neurotransmitters. pumps cytosolic monoamines including dopamine, norepinephrine, serotonin, and histamine into synaptic vesicles. It is a requisite for vesicular amine storage prior to secretion via exocytosis.
Specifications

Specifications

VMAT2
Polyclonal
Unconjugated
SLC18A2
Liquid
SLC18A2
Reacts with an internal sequence of the human, mouse and rat VMAT2 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rabbit, Rat
Immunohistochemistry (Frozen), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q05940, Q8BRU6, Q01827
SVAT, SVMT, VAT2, VMAT2, monoamine neurotransmitter transporter, monoamine transporter, solute carrier family 18 member 2, synaptic vesicular amine transporter, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, vesicular amine transporter 2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100351780, 214084, 25549, 6571
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit