Learn More
Vomeronasal 1 Receptor A1 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00244W
Catalog No. OSR00244W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Putative pheromone receptor implicated in the regulation of social as well as reproductive behavior. Expressed in a subset of sensory neurons located in the apical layer of the vomeronasal organ. Mice lacking all but one V1RA and V1RB gene (12% of the V1R repertoire) show a lack of chemosensory response to a subset of known pheromonal ligands and changes in maternal aggression as well as male reproductive behavior.
Specifications
Specifications
Vomeronasal 1 Receptor A1
Polyclonal
whole serum with no preservative
Q8VIC6
Vomeronasal type-1 receptor A8, pheromone receptor 1, VN1A1, mV1R1, V1ra1, Pr1, V1ra8, V1R, V1r1, VN12
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Vmn1r51
Lyophilized
VMN1R51
Synthetic peptide from mouse vomeronasal type-1 receptor A1.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22296