Learn More
Vomeronasal 1 Receptor A3/A4 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00240W
Catalog No. PIOSR00240W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Putative pheromone receptor implicated in the regulation of social and reproductive behavior. Mice lacking all but one V1RA and V1RB gene (12% of the V1R repertoire) show a lack of chemosensory response to a subset of known pheromonal ligands and changes in maternal aggression as well as male reproductive behavior.
Specifications
Specifications
Vomeronasal 1 Receptor A3/A4
Polyclonal
whole serum with no preservative
Q9EQ51, Q9EQ52
Vmn1r47, Vmn1r48, Vomeronasal type-1 receptor A6, VN1A3, V1ra3, V1ra6, V1RA1, V1RA3, vomeronasal type-1 receptor A7, VN1A4, VN3, V1ra4, V1ra7
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Vmn1r48
Lyophilized
VMN1R47, VMN1R48
Synthetic peptide from mouse vomeronasal type-1 receptor A3 & A4.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
113845, 113846