Learn More
Vomeronasal 1 Receptor B1 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00237W
Catalog No. PIOSR00237W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Putative pheromone receptor implicated in the regulation of social and reproductive behavior. Expressed in a subset of sensory neurons located in the apical layer of the vomeronasal organ. Mice lacking all but one V1RA and V1RB gene (12% of the V1R repertoire) show a lack of chemosensory response to a subset of known pheromonal ligands and changes in maternal aggression as well as male reproductive behavior.
Specifications
Specifications
Vomeronasal 1 Receptor B1
Polyclonal
whole serum with no preservative
Q9EP51
VN1B1, V1rb1, V1ra5, VN2, V1RA5, vomeronasal type-1 receptor A5, vomeronasal receptor 2, pheromone receptor VN2, MGC130072
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Vmn1r50
Lyophilized
VMN1R50
Synthetic peptide from mouse vomeronasal type-1 receptor B1.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
113852