Learn More
Vomeronasal 1 Receptor B7/A11 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00232W
Catalog No. PIOSR00232W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Putative pheromone receptor implicated in the regulation of social and reproductive behavior. Mice lacking all but one V1RA and V1RB gene (12% of the V1R repertoire) show a lack of chemosensory response to a subset of known pheromonal ligands and changes in maternal aggression as well as male reproductive behavior.
Specifications
Specifications
Vomeronasal 1 Receptor B7/A11
Polyclonal
whole serum with no preservative
Q9EQ46
Vomeronasal type-1 receptor A11, V1rb7, V1ra11, VN1B7
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Vmn1r40
Lyophilized
VMN1R40
Synthetic peptide from mouse vomeronasal type-1 receptor B7 or A11.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
113855