Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Von Hippel Lindau, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide derived from the human Von Hippel Lindau protein. [UniProt# P40337]
Antigen Von Hippel Lindau
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001899SS
View Documents
Novus Biologicals
NB1001899SS
0.025 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Von Hippel Lindau Polyclonal antibody specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A synthetic peptide derived from the human Von Hippel Lindau protein. [UniProt# P40337]
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
P40337
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
IgG
Von Hippel Lindau
Affinity Purified
RUO
Human. Not expected to work in mouse based on sequence divergence.
7428
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit