Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPARP Mouse anti-Human, Clone: P193-6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135747

Catalog No. 01-672-053


Add to cart

Description

Description

This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.
Specifications

Specifications

VPARP
Monoclonal
0.25 mg/mL
antibody with 1% BSA and 0.1% sodium azide
Q9UKK3
ADPRTL1, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193
Mouse
IgG2b, kappa
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
143
Immunohistochemistry (Frozen), Western Blot
P193-6
Unconjugated
PARP4
Liquid
PARP4
Proprietary immunogen.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit