Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521854

Catalog No. PIPA521854


Add to cart

Description

Description

PA5-21854 targets VPS11 in WB applications and shows reactivity with Human samples. The PA5-21854 immunogen is a synthetic peptide corresponding to a region within amino acids 300 and 392 of VPS11 (Uniprot IDNo.Q9H270). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

VPS11
Polyclonal
Unconjugated
VPS11
Liquid
VPS11
Synthetic peptide corresponding to a region within amino acids 300 and 392 of VPS11 (Uniprot IDNo.Q9H270)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9H270
PP3476, END1, PEP5, RNF108, hVPS11, RING finger protein 108, vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55823
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit