Learn More
anti-VPS24, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130773
Catalog No. NBP130773
Add to cart
Description
Description
VPS24 Polyclonal antibody specifically detects VPS24 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
VPS24
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CHMP3
A synthetic peptide from the n-terminal of human CHMP3 (Charged multivesicular body protein 3, Vacuolar protein sorting associated protein 24, VPS24) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat and mouse.
IgG purified
RUO
Primary
51652
Reconstitute with 0.5 ml sterilized water.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Sterile water with No Preservative
CGI-149, CHMP325.1 protein, Chromatin-modifying protein 3, hVps24, member 3,25.1, NEDFFLJ38114, vacuolar protein sorting 24 (yeast), vacuolar protein sorting 24 homolog (S. cerevisiae)
Sheep
IgG
0.5 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for CHMP3.
Human, Mouse, Rat