Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS26A Goat anti-Bovine, Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA132102

Catalog No. PIPA132102


Add to cart

Description

Description

PA1-32102 detects VPS26 from human samples. PA1-32102 has been successfully used in Western blot applications. The PA1-32102 immunogen is: Peptide with sequence SPESQASAEQPEM, from C Terminus of the protein sequence according to NP_004887 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

VPS26A
Polyclonal
Unconjugated
VPS26A
Liquid
VPS26A
Peptide with sequence SPESQASAEQPEM, from C Terminus of the protein sequence according to NP_004887
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Porcine, Rat
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q0VD53, O75436, P40336, Q6AY86
FLJ12930, HB58, Hbeta58, PEP8A, VPS26
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100155086, 30930, 361846, 510574, 9559
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit