Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS29 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528117

Catalog No. PIPA528117


Add to cart

Description

Description

PA5-28117 targets VPS29 in WB applications and shows reactivity with Human samples. The PA5-28117 immunogen is synthetic peptide corresponding to a region within amino acids 120 and 182 of Human VPS29. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene belongs to a group of vacuolar protein sorting (VPS) genes that, when functionally impaired, disrupt the efficient delivery of vacuolar hydrolases. The protein encoded by this gene is a component of a large multimeric complex, termed the retromer complex, which is involved in retrograde transport of proteins from endosomes to the trans-Golgi network. This VPS protein may be involved in the formation of the inner shell of the retromer coat for retrograde vesicles leaving the prevacuolar compartment. Alternative splice variants encoding different isoforms, and usage of multiple polyadenylation sites have been found for this gene.
Specifications

Specifications

VPS29
Polyclonal
Unconjugated
VPS29
Liquid
VPS29
Synthetic peptide corresponding to a region within amino acids 120 and 182 of Human VPS29
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q9UBQ0
DC15, PEP11, PEP11 homolog, hVPS29, retromer protein, vacuolar protein sorting-associated protein 29, vacuolar sorting protein VPS29/PEP11, vesicle protein sorting 29, x 007 protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51699
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit