Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG2 Mouse anti-Human, Clone: 541527, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524242

Catalog No. PIMA524242


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human VSIG1, 3, 4, or recombit mouse VSIG2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

VSIG2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q96IQ7
CTH, CTXL, 2210413P10Rik, Cortical thymocyte-like protein, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23584
Western Blot
541527
Unconjugated
VSIG2
Lyophilized
VSIG2
Mouse myeloma cell line NS0-derived recombit human VSIG2 Val24-Ala243
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit