Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-G Tag Goat anti-Tag, FITC, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131803

Catalog No. PA131803

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

PA1-31805 detects RNF3 from human samples. PA1-31805 has been successfully used in Western blot applications. The PA1-31805 immunogen is: Peptide with sequence PLLLHYRPKMDLL, from C Terminus of the protein sequence according to NP_006306.

Vesicular stomatitis virus (VSV), an enveloped RNA virus from the Rhabdoviridae family, is released from the plasma membrane of host cells by a process called budding. The glycoprotein (VSV G) contains a domain in its extracellular membrane-proximal stem that appears to be needed for efficient VSV budding: Sequence: YTDIEMNRLGK.
Specifications

Specifications

VSV-G Tag
Polyclonal
FITC
Liquid
Goat
IgG
100 μg
4° C, do not freeze
Primary
Tag
Immunocytochemistry, Immunofluorescence
1 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide; pH 7.2
Vesicular stomatitis virus; VSVG tag; VSV tag; VSV-G epitope tag; VSVG epitope tag; VSIVgp3; VSV epitope tag; YTDIEMNRLGK
synthetic peptide YTDIEMNRLGK conjugated to KLH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit