Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-G Tag Rabbit anti-Tag, HRP, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126564

Catalog No. PA126564


Add to cart

Description

Description

PA1-26564 detects VSV-G tag. PA1-26564 has been successfully used in Western blot and ELISA procedures. The PA1-26564 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues Y T D I E M N R L G K of VSV-G tag.

Vesicular stomatitis virus (VSV), an enveloped RNA virus from the Rhabdoviridae family, is released from the plasma membrane of host cells by a process called budding. The glycoprotein (VSV G) contains a domain in its extracellular membrane-proximal stem that appears to be needed for efficient VSV budding: Sequence: YTDIEMNRLGK.
Specifications

Specifications

VSV-G Tag
Polyclonal
HRP
Liquid
Rabbit
IgG
100 μg
4° C
Primary
Tag
ELISA, Western Blot
1 mg/mL
HEPES buffered saline with 0.2% BSA and 0.002% thimerosal; pH 7.0
Vesicular stomatitis virus; VSVG tag; VSV tag; VSV-G epitope tag; VSVG epitope tag; VSIVgp3; VSV epitope tag; YTDIEMNRLGK
Synthetic peptide: YTDIEMNRLGK conjugated to KLH
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit