Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vti1b Mouse, Unlabeled, Clone: 7, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611405

 View more versions of this product

Catalog No. BDB611405


Add to cart

Description

Description

Eukaryotic protein trafficking involves packaging of target molecules into membranous vesicles that bud from a donor compartment, travel to a specific destination, fuse, and release their contents into an acceptor compartment. Recognition between vesicle and acceptor membrane is mediated by the pairing of the integral membrane SNARE proteins. The stable interaction between vesicle proteins (v-SNAREs; VAMP1, VAMP2) and target proteins (t-SNAREs; syntaxin 1, SNAP-25) juxtaposes the membranes and results in an activated docked state and/or membrane fusion. With the identification of all SNARE family members in yeast, the research focus has turned to mammalian cells. Here, sequence analysis has identified additional SNARE proteins, including VTI1a and VTI1b. In line with their involvement in vesicle transport, these molecules are expressed in a wide range of mammalian tissues. VTI1b is a membrane bound protein whose localization overlaps with the cis/medial Golgi marker mannosidase II and the trans-Golgi marker syntaxin 6. VTI1b interacts with, and disrupts the localization of, syntaxin 5. Thus, VTI1b is thought to function in the regulation of post-Golgi vesicle trafficking.

Host Species: Mouse
Clone: 7
Isotype: IgG1
Species Reactivity: Human
Immunogen: Mouse Vti1b aa. 9-121
Formula Weight [Chemical]: 27kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

Vti1b
7
Unconjugated
Affinity Purified
Mouse Vti1b aa. 9-121
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.