Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VTI1B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA518502
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
VTI1B Polyclonal antibody specifically detects VTI1B in Human samples. It is validated for Western Blot.Specifications
VTI1B | |
Polyclonal | |
Unconjugated | |
Vti1b | |
A novel SNARE; AU015348; GES30; HGNC:17793; MVti1b; SNARE; Soluble N-ethylmaleimide-sensitive factor-attachment protein receptor; vesicle transport through interaction with t-SNAREs 1B; vesicle transport through interaction with t-SNAREs 1B homolog; vesicle transport through interaction with t-SNAREs homolog 1B; Vesicle transport v-SNARE protein Vti1-like 1; vesicle-associated soluble NSF attachment protein receptor; v-SNARE; VTI1; VTI1B; VTI1L; VTI1L1; VTI1-LIKE; vti1-rp1; VTI2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10490 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UEU0 | |
Vti1b | |
Synthetic peptide sequence (RDFDEKQQEANET) corresponding to the internal amino acids of VTI1B | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction