Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitronectin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525964

Catalog No. PIPA525964

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
Specifications

Specifications

Vitronectin
Polyclonal
PBS with 0.09% sodium azide
P04004
V75, VN, VNT, S-protein, complement S-protein, epibolin, serum spreading factor, serum-spreading factor, somatomedin B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7448
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
VTN
Liquid
VTN
KLH conjugated synthetic peptide between 65-93 amino acids from the N-terminal region of human VTN
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit