Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VWA3B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523810

Catalog No. PIPA523810

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

VWA3B
Polyclonal
PBS with 0.09% sodium azide
Q502W6
von Willebrand factor A domain-containing protein 3B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
200403, 70853
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
VWA3B
Liquid
VWA3B
KLH conjugated synthetic peptide between 1178-1207 amino acids from the C-terminal region of human VWA3B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit