Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-WDR24, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$366.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Recombinant protein encompassing a sequence within the center region of human WDR24. The exact sequence is proprietary.
Antigen WDR24
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP220892
View Documents
Novus Biologicals
NBP220892
0.1 ml
Each for $366.50
Add to cart
 
NBP220892SS
View Documents
Novus Biologicals
NBP220892SS
0.02 ml This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

WDR24 Polyclonal antibody specifically detects WDR24 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
Recombinant protein encompassing a sequence within the center region of human WDR24. The exact sequence is proprietary.
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
84219
0.96mg/mL
IgG
WDR24
Affinity Purified
RUO
Q96S15
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit