Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126630
Description
WDR26 Polyclonal specifically detects WDR26 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).Specifications
WDR26 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
MIP2, myocardial ischemic preconditioning up-regulated protein 2, WD repeat domain 26, WD repeat-containing protein 26 | |
Rabbit | |
Affinity Purified | |
RUO | |
80232 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation (Negative) | |
BAG10986.1 | |
WDR26 | |
The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human WD repeat domain 26 using the numbering given in entry BAG10986.1 (GeneID 80232). | |
100 μL | |
Primary | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction