Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR4, Polyclonal, Invitrogen™

Catalog No. PIPA127801
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127801 50 μg
1 options

Catalog No. PIPA127801

Supplier: Thermo Scientific PA127801

Chicken Polyclonal Antibody

Required for 7-methylguanosine modification of tRNA. This protein is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD). which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes. including cell cycle progression. signal transduction. apoptosis. and gene regulation. This protein is excluded as a candidate for a form of nonsyndromic deafness (DFNB10). but is still a candidate for other disorders mapped to 21q22.3 as well as for the development of Down syndrome phenotypes. Alternative splicing of this gene generates at least 3 transcript variants. two of which diverge at the 3' UTR and thus encode the same protein products.

PA1-27801 detects WDR4 from human samples.PA1-27801 may be successfully used in Western blot applications.The immunogen is a synthetic peptide corresponding to human WD repeat protein 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR4
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene WDR4
Gene Accession No. P57081
Gene Alias TRM82
Gene Symbols WDR4
Host Species Chicken
Immunogen Synthetic peptide corresponding to human WD repeat protein 4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10785
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.