Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-WDR57, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192586

Catalog No. NBP192586


Add to cart

Description

Description

WDR57 Polyclonal antibody specifically detects WDR57 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

WDR57
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
38 kDa-splicing factor, HPRP8BP, Prp8-binding protein, PRP8BP40K, PRPF8BPFLJ41108, RP11-490K7.3, SFP38, small nuclear ribonucleoprotein 40kDa (U5), SPF38MGC1910, U5 small nuclear ribonucleoprotein 40 kDa protein, U5 snRNP 40 kDa protein, U5 snRNP-specific 40 kDa protein, U5 snRNP-specific 40 kDa protein (hPrp8-binding), U5-40K, U5-40kD protein, WD repeat domain 57 (U5 snRNP specific), WD repeat-containing protein 57, WDR57
Rabbit
IgG
0.1 ml
Core ESC Like Genes, Stem Cell Markers
Primary
9410
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Knockdown
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Affinity Purified
SNRNP40
This antibody was developed against Recombinant Protein corresponding to amino acids:GDCDNYATLKGHSGAVMELHYNTDGSMLFSASTDKTVAVWDSETGERVKRLKGHTSFVNSCYPARRGPQLVCTGSDDGTVKLWDIRKKAAIQ
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only