Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THO complex 6 homolog (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human WDR58 protein.
Supplier: Abnova Corporation H00079228B01
Description
Sequence: MTIFSQSVSPCGKFLAAGNNYGQIAIFSLSSALSSEAKEESKKPVVTFQAHDGPVYSMVSTDRHLLSAGDGEVKAWLWAEMLKKGCKELWRRQPPYRTSLEVPEINALLLVPKENSLILAGGDCQLHTMDLETGTFTRVLRGHTDYIHCLALRERSPEVLSGGEDGAVRLWDLRTAKEVQTIEVYKHEECSRPHNGRWIGCLATDSDWMVCGGGPALTLWHLRSSTPTTIFPIRAPQKHVTFYQDLILSAGQGRCVNQWQLSGELKAQVPGSSPGLLSLSLNQQPAAPECKVLTAAGNSCRVDVFTNLGYRAFSLSFSpecifications
THO complex 6 homolog (Drosophila) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human WDR58 protein. | |
THOC6 | |
MGC2655/WDR58/fSAP35 | |
Mouse | |
50 μL | |
Kinases | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC003118 | |
THOC6 | |
WDR58 (AAH03118, 1 a.a. ∼ 317 a.a) full-length human protein. | |
RUO | |
Yes | |
79228 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction