Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524529

Catalog No. PIPA524529

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This intronless gene encodes a protein containing several WD40 repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, including a trp-asp at the C-terminal end. The encoded protein may mediate protein-protein interactions.
Specifications

Specifications

WDR5B
Polyclonal
PBS with 0.09% sodium azide
Q86VZ2
WD repeat-containing protein 5B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54554
Flow Cytometry, Western Blot
Unconjugated
WDR5B
Liquid
WDR5B
KLH conjugated synthetic peptide between 58-85 amino acids from the N-terminal region of human WDR5B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit