Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR76 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524147

Catalog No. PIPA524147


Add to cart

Description

Description

PA5-24147 detects WDR76 in WB application.

Specifications

Specifications

WDR76
Polyclonal
Unconjugated
WDR76
Liquid
WDR76
KLH conjugated synthetic peptide between 455-484 amino acids from the C-terminal region of human WDR76
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9H967, A6PWY4
CDW14, WD repeat-containing protein 76
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
241627, 79968
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit