Learn More
WD repeat domain 77, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human WDR77 protein.
Supplier: Abnova Corporation H00079084B01
Description
WDR77 is a component of the 20S PRMT5 (MIM 604045)-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins (see MIM 601061). This modification targets Sm proteins to the survival of motor neurons (SMN) complex (see MIM 600354) for assembly into small nuclear ribonucleoprotein core particles (Friesen et al., 2002 [PubMed 11756452]).[supplied by OMIM
Sequence: MRKETPPPLVPPAAREWNLPPNAPACMERQLEAARYRSDGALLLGASSLSGRCWAGSLWLFKDPCAAPNEGFCSAGVQTEAGVADLTWVGERGILVASDSGAVELWELDENETLIVSKFCKYEHDDIVSTVSVLSSGTQAVSGSKDICIKVWDLAQQVVLSSYRAHAAQVTCVAASPHKDSVFLSCSEDNRILLWDTRCPKPASQIGCSAPGYLPTSLAWHPQQSEVFVFGDENGTVSLVDTKSTSCVLSSAVHSQCVTGLVFSPHSVPFLASLSEDCSLAVLDSSLSELFRSQAHRDFVRDATWSPLNHSLLTTVGWDHQVVHHVVPTEPLPAPGPASVTESpecifications
WD repeat domain 77 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human WDR77 protein. | |
WDR77 | |
HKMT1069/MEP50/MGC2722/Nbla10071/RP11-552M11.3/p44 | |
Mouse | |
50 μL | |
Yes | |
79084 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_024102.2 | |
WDR77 | |
WDR77 (NP_077007.1, 1 a.a. ∼ 342 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.