Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR77 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA519806

Catalog No. PIPA519806

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rhesus monkey and orangutan based on sequence homology.

WDR77 is a component of the 20S PRMT5 complex for assembly into small nuclear ribonucleoprotein core particles .
Specifications

Specifications

WDR77
Polyclonal
Unconjugated
WDR77
Liquid
WDR77
Synthetic peptide conjugated to KLH derived from within residues 300 to the C-terminus of Human WDR77.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Q9BQA1
HKMT1069, MEP-50, MEP50, MGC2722, Nbla10071, RP11-552M11.3, p44, p44/Mep50, WD repeat-containing protein 77; androgen receptor cofactor p44; methylosome protein 50
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
79084
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit