Learn More
WDR82 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524741
Catalog No. PIPA524741
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
WDR82 is a component of the mammalian SET1A (MIM 611052)/SET1B (MIM 611055) histone H3-Lys4 methyltransferase complexes.
Specifications
Specifications
WDR82
Polyclonal
Unconjugated
WDR82
Liquid
WDR82
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human WDR82
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6UXN9
MST107, MSTP107, PRO2730, PRO34047, SWD2, TMEM113, WDR82A, WD repeat-containing protein 82, transmembrane protein 113
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80335