Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDSOF1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524324

Catalog No. PIPA524324

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Possible role in ribosomal RNA processing. May function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
Specifications

Specifications

WDSOF1
Polyclonal
Unconjugated
DCAF13
Liquid
DCAF13
KLH conjugated synthetic peptide between 369-398 amino acids from the C-terminal region of human WDSOF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9NV06, Q6PAC3
GM83, WDSOF1, DDB1- and CUL4-associated factor 13, WD repeat and SOF domain-containing protein 1, WD repeats and SOF1 domain containing
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
223499, 25879
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit