Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDYHV1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512414

Catalog No. PIPA512414

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

WDYHV1
Polyclonal
PBS with 0.09% sodium azide
Q96HA8, Q80WB5
WDYHV motif-containing protein 1; WDYHV1; C8orf32
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55093, 76773
Immunohistochemistry, Western Blot
Unconjugated
WDYHV1
Liquid
WDYHV1
KLH conjugated synthetic peptide between 106-136 amino acids from the central region of human WDYHV1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit