Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRN Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527320

Catalog No. PIPA527320


Add to cart

Description

Description

PA5-27320 targets Werner syndrome protein in IF and WB applications and shows reactivity with Human samples. The PA5-27320 immunogen is recombit fragment corresponding to a region within amino acids 1194 and 1432 of Human Werner syndrome protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the RecQ subfamily and the DEAH (Asp-Glu-Ala-His) subfamily of DNA and RNA helicases. DNA helicases are involved in many aspects of DNA metabolism, including transcription, replication, recombination, and repair. This protein contains a nuclear localization signal in the C-terminus and shows a predomit nucleolar localization. It possesses an intrinsic 3 ft. to 5 ft. DNA helicase activity, and is also a 3 ft. to 5 ft. exonuclease. Based on interactions between this protein and Ku70/80 heterodimer in DNA end processing, this protein may be involved in the repair of double strand DNA breaks. Defects in this gene are the cause of Werner syndrome, an autosomal recessive disorder characterized by premature aging.
Specifications

Specifications

WRN
Polyclonal
Unconjugated
WRN
Liquid
WRN
Recombit fragment corresponding to a region within amino acids 1194 and 1432 of Human Werner syndrome protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q14191
RECQ3, RECQL2, RECQL3, DNA helicase, RecQ-like type 3, Werner syndrome ATP-dependent helicase, exonuclease WRN, recQ protein-like 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7486
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit