Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

WT1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138865

Catalog No. 01-674-490



Description

Description

This antibody is predicted to react with canine, mouse, porcine, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is Wilm ft.s tumor tissue.

Wilm’s Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm’s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelioma.
Specifications

Specifications

WT1
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P22561, P19544
WT, AWT1, EWS-WT1, GUD, NPHS4, WAGR, WIT-2, WT33, Wilms tumor protein, amino-terminal domain of EWS, last three zinc fingers of the DNA-binding domain of WT1
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7490, 22431
Immunohistochemistry (Paraffin)
Unconjugated
WT1
Liquid
WT1
Synthetic peptide from the C-terminus of human Wilm′s Tumor protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit