Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIPF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524457

Catalog No. PIPA524457


Add to cart

Description

Description

This gene encodes a WASP interacting protein (WIP)-related protein. It has been shown that this protein has a role in the WASP-mediated organization of the actin cytoskeleton and that this protein is a potential link between the activated platelet-derived growth factor receptor and the actin polymerization machinery.
Specifications

Specifications

WIPF2
Polyclonal
PBS with 0.09% sodium azide
Q8TF74
WICH, WIRE, WAS/WASL-interacting protein family member 2, WASP-binding protein, WASP-interacting protein-related protein, WIP- and CR16-homologous protein, WIP-related protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
147179
Immunohistochemistry, Western Blot
Unconjugated
WIPF2
Liquid
WIPF2
KLH conjugated synthetic peptide between 392-419 amino acids from the C-terminal region of human WIPF2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit