Learn More
MilliporeSigma™ anti-WIPI1 Clone: 3C1, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1505100UG
Catalog No. ST1505100UG
Add to cart
Description
Description
Specifically detects WIPI1 Clone: 3C1 in Human, Rat samples, and it is validated for Immuno Blotting
WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a β-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI1, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
WIPI1
3C1
In PBS, pH 7.2.
Murine
IgG2b
RUO
Primary
Recognizes the ∽45kDa WIPI1 protein in PC-12 cells.
Immuno Blotting, Immunoblot
Unconjugated
Purified
A recombinant polypeptide corresponding to amino acids 348-446 of human WIPI1, expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human, Rat