Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WNK1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of WNK1.
Manufacturer: Abnova Corporation PAB16446
Description
The WNK1 gene encodes a cytoplasmic serine-threonine kinase expressed in distal nephron.[supplied by OMIM
Specifications
WNK1 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (7 ug/mL) The optimal working dilution should be determined by the end user. | |
WNK1 | |
KDP|KIAA0344|PHA2C|PRKWNK1|PSK | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
Primary | |
65125 | |
Human, Monkey |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of WNK1. | |
In PBS (0.09% sodium azide) | |
Liquid | |
WNK1 | |
A synthetic peptide (conjugated with KLH) corresponding to human WNK1. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Internal domain of human. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only