anti-WNK1, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 3646000201MG

 View more versions of this product

Catalog No. 36-460-00201

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Disclaimers:

For Research Use Only


    Specifications

    Host Species Rabbit
    Applications ELISA,Western Blotting
    Quantity 0.1mg
    Antigen WNK1
    Format Affinity Purified
    Gene Accession No. Q6IFS5
    Immunogen This antibody is specific for the Middle Region (aa 80-230) of the target protein
    Gene ID (Entrez) 65125
    Cross Reactivity Human
    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Test Specificity This product is specific for Human WNK1.
    Conjugate Unlabeled

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit