Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-WNK3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP103340

Catalog No. NBP103340


Add to cart

Description

Description

WNK3 Polyclonal specifically detects WNK3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

WNK3
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
EC 2.7.11, FLJ30437, KIAA1566FLJ42662, PRKWNK3lysine deficient 3, WNK lysine deficient protein kinase 3
Rabbit
198 kDa
0.1 mL
Primary
Human
IgG
Western Blot, Immunoprecipitation
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Q9BYP7
WNK3
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human WNK lysine deficient protein kinase 3 using the numbering given in entry NP_001002838.1 (GeneID 65267).
Affinity Purified
RUO
65267
Store at 4C. Do not freeze.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only