Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Wnt-4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145930

Catalog No. NBP145930


Add to cart

Description

Description

Wnt-4 Polyclonal antibody specifically detects Wnt-4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Wnt-4
0.5 mg/mL
Western Blot 1 - 4 ug/mL
BAC23080
protein Wnt-4, SERKAL, wingless-type MMTV integration site family, member 4, WNT-4
Rabbit
IgG
0.1 mg
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary
54361
Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Affinity Purified
WNT4
Synthetic peptide surrounding amino acid 10 of human Wnt-4.
Affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only