Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Wnt-6, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145931

Catalog No. NBP145931

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Wnt-6
0.5 mg/mL
Western Blot
NP_033552.2
protein Wnt-6, wingless-type MMTV integration site family, member 6
Rabbit
IgG
0.1 mg
Signal Transduction, Wnt Signaling Pathway
Primary
7475
Western Blot
Unconjugated
PBS, 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
WNT6
Synthetic peptide surrounding amino acid 276 of mouse Wnt-6.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only