Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WNT9A Mouse anti-Human, APC, Clone: 326711, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523637

Catalog No. PIMA523637


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human Wnt-2, -3a, -6, -7a, -7b, recombit mouse Wnt-1, -2b, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, or -16 is observed.

Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
Specifications

Specifications

WNT9A
Monoclonal
APC
WNT9A
Liquid
WNT9A
E. coli-derived recombit human Wnt-9a Pro48-Glu92 and Arg231-Phe304
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
326711
Buffered saline solution with BSA and 0.1% sodium azide
O14904
WNT9A, Protein Wnt-14, Protein Wnt-9a, WNT14
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
7483
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit