Learn More
WNT2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WNT2.
Supplier: Abnova Corporation PAB9583
Description
This gene is a member of the WNT gene family. The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Alternatively spliced transcript variants have been identified for this gene. [provided by RefSeq
Specifications
WNT2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of WNT2. | |
In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
INT1L1/IRP | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes 34 KDa Wnt-2. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
WNT2 | |
WNT2 | |
A synthetic peptide corresponding to residues surrounding amino acids 45 of human WNT2. | |
RUO | |
7472 | |
Human, Mouse, Rat | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.