Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WNT5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA534325
Description
Percent identity with other species by BLAST analysis: Human, Orangutan, Gibbon, Monkey, Marmoset (100%) Gorilla, Elephant, Panda, Dog, Horse, Rabbit, Pig, Opossum (93%).
WNT5B Polyclonal antibody specifically detects WNT5B in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
WNT5B | |
Polyclonal | |
Unconjugated | |
WNT5B | |
AW545702; protein Wnt-5b; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5B; Wnt family member 5B; WNT5B; Wnt-5b; WNT-5B protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81029 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H1J7 | |
WNT5B | |
Synthetic 14 amino acid peptide from internal region of human WNT5B. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction