Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XAB2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527101

Catalog No. PIPA527101


Add to cart

Description

Description

PA5-27101 targets XAB2 in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-27101 immunogen is recombit fragment corresponding to a region within amino acids 30 and 243 of Human XAB2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Involved in transcription-coupled repair (TCR), transcription and pre-mRNA splicing.
Specifications

Specifications

XAB2
Polyclonal
Unconjugated
XAB2
Liquid
XAB2
Recombit fragment corresponding to a region within amino acids 30 and 243 of Human XAB2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1.01 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9HCS7, Q9DCD2
HCNP, HCRN, NTC90, SYF1, SYF1 homolog, RNA splicing factor, XPA-binding protein 2, crn-related protein kim1, pre-mRNA-splicing factor SYF1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
56949, 67439
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit