Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
$204.00 - $409.50
Specifications
Antigen | XAF1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056355T
|
Novus Biologicals
NB10056355SS |
0.025 mg |
Each for $204.00
|
|
|||||
NB10056355
|
Novus Biologicals
NB10056355 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
XAF1 Polyclonal specifically detects XAF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
XAF1 | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
BIRC4-binding protein, BIRC4BPBIRC4 binding protein, HSXIAPAF1, XIAP associated factor 1, XIAPAF1XIAP-associated factor 1 | |
XAF1 | |
IgG | |
Protein G purified | |
In human liver, a 34 kDa band should be observed. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
54739 | |
Amino acids 201-218 (NQTSTMEKDVRPKTRSIN) and 301-317 (QLDLEKERYYKFKRFHF) of human XAF1, GenBank no. CAA68030.1, were used as immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title