Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-XBP1, Alexa Fluor 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177681AF647

Catalog No. NBP177681Y


Add to cart

Description

Description

XBP1 Polyclonal antibody specifically detects XBP1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

XBP1
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
XBP1
A genomic peptide made to an internal region of the human XBP1 protein (within residues 100-250). [Swiss-Prot P17861]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
This antibody is specific for both XBP1s and XBP1u.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
P17861
TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5
Rabbit
IgG
0.1 ml
Cancer, Lipid and Metabolism, Unfolded Protein Response
Primary
7494
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only