Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-XBP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications

Manufacturer:  Novus Biologicals NBP177681

 View more versions of this product

Catalog No. NBP177681


Add to cart

Description

Description

XBP1 Polyclonal specifically detects XBP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

XBP1
0.58 mg/mL
Western Blot 1 ug/mL, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100
P17861, O35426
TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP1s, XBP1u, XBP2Tax-responsive element-binding protein 5
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for both XBP1s and XBP1u.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS and 30% Glycerol with 0.05% Sodium Azide
Affinity Purified
XBP1
A genomic peptide made to an internal region of the human XBP1 protein (within residues 100-250). [Swiss-Prot P17861]
Immunogen affinity purified
RUO
Primary
7494
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only