Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XBP1, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127594
Description
Transcription factor essential for hepatocyte growth. the differentiation of plasma cells. the immunoglobulin secretion. and the unfolded protein response (UPR). Acts during endoplasmic reticulum stress (ER) by activating unfolded protein response (UPR) t
PA1-27594 detects XBP1 from human samples.PA1-27594 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human X box-binding protein 1.Specifications
XBP1 | |
Polyclonal | |
XBP1 | |
TREB5, XBP2 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
7494 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
P17861 | |
XBP1 | |
Synthetic peptide corresponding to human X box-binding protein 1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction