Learn More
XBP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525010
Catalog No. PIPA525010
Add to cart
Description
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
XBP1 encodes a transcription factor that regulates MHC class II genes by binding to a promoter element referred to as an X box. This gene product is a bZIP protein, which was also identified as a cellular transcription factor that binds to an enhancer in the promoter of the T cell leukemia virus type 1 promoter. It may increase expression of viral proteins by acting as the DNA binding partner of a viral transactivator. It has been found that upon accumulation of unfolded proteins in the endoplasmic reticulum (ER), the mRNA of this gene is processed to an active form by an unconventional splicing mechanism that is mediated by the endonuclease inositol-requiring enzyme 1 (IRE1). The resulting loss of 26 nt from the spliced mRNA causes a frame-shift and an isoform XBP1(S), which is the functionally active transcription factor.
Specifications
Specifications
XBP1
Polyclonal
Unconjugated
XBP1
Liquid
XBP1
KLH conjugated synthetic peptide between 62-91 amino acids from the central region of human XBP1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P17861, O35426
TREB5, XBP2, X-box-binding protein 1, XBP-1, tax-responsive element-binding protein 5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22433, 7494