Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse XCL1/Lymphotactin Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems MAB486
This item is not returnable.
View return policy
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
XCL1/Lymphotactin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6375 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, ELISA | |
80222 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
AAA56752 | |
XCL1 | |
E. coli-derived recombinant mouse XCL1/Lymphotactin Val22-Gly114 Accession # AAA56752 | |
500 μg | |
Primary | |
Detects mouse XCL1/Lymphotactin in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human XCL1/Lymphotactin is observed and no cross-reactivity with recombinant viral (rv) CMV UL147 is observed. In Western blots, 75% cross-reactivity with rvCMV UL147 is observed and no cross-reactivity with recombinant human CCL1-3, 5, 7, 8, 11, 13-25, 28, recombinant mouse CCL1-7, 9, 11, 12, 15, 19, 21, 22, 24, 25, 27 or 28 is observed. | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction